Learn More
Perkin Elmer US LLC Demountable Quartz Injector, 1.5 mm I.D.
Catalog No. 509045598
Encompass_Preferred
This Quartz Injector Cassette Mount is designed for use with demountable torches in NexION 300/350 ICP-MS series instruments. The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch. Standard Injectors have an O-ring free direct connection to Cyclonic and PFA spray chambers while O-ring free Injectors have an O-ring free seal that reduces contamination, makes it easy to clean and easy to insert.
Catalog No. 50-904-5598 Supplier Perkin Elmer US LLC Supplier No. N8122413
This Quartz Injector Cassette Mount is designed for use with demountable torches in NexION 300/350 ICP-MS series instruments. The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch. Standard Injectors have an O-ring free direct connection to Cyclonic and PFA spray chambers while O-ring free Injectors have an O-ring free seal that reduces contamination, makes it easy to clean and easy to insert.