Learn More
Perkin Elmer US LLC 1.2 mm I.D. Alumina Injector for Optima 4300V/5300V/7300V
Catalog No. 509043814
Encompass_Preferred
This Alumina Injector is included in the Quick-Change Torch Module for Oil Analysis (N0770910) and the Organics Sample Introduction Kit for Optima 4300 V/5300 V/7300 V (N0770732).
Catalog No. 50-904-3814 Supplier Perkin Elmer US LLC Supplier No. N0771531
This Alumina Injector is included in the Quick-Change Torch Module for Oil Analysis (N0770910) and the Organics Sample Introduction Kit for Optima 4300 V/5300 V/7300 V (N0770732).