Learn More
Glass Expansion Injector Adaptor Avio 200/500
SDP
Catalog No. 502703117
Encompass_Preferred
Injector Adaptor for D-Torch. Compatible with Glass Expansion D-Torch for PerkinElmer Avio 200/500 ICP-OES.Replacement O-Ring Kit P/N 31-808-4077.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3117 Supplier Glass Expansion Supplier No. 318083782
Injector Adaptor for D-Torch. Compatible with Glass Expansion D-Torch for PerkinElmer Avio 200/500 ICP-OES.Replacement O-Ring Kit P/N 31-808-4077.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001