Learn More
Glass Expansion Injector Adaptor for D-Torch Spectro SOP
SDP
Catalog No. 502703143
Encompass_Preferred
Injector Adaptor for D-Torch. Compatible with Glass Expansion D-Torch (P/N 501907023, 30-808-2928) for Spectro (Ametek) SOP Modula, Ciros, Genesis, and Arcos Radial ICP-OES. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3143 Supplier Glass Expansion Supplier No. 318083177
Injector Adaptor for D-Torch. Compatible with Glass Expansion D-Torch (P/N 501907023, 30-808-2928) for Spectro (Ametek) SOP Modula, Ciros, Genesis, and Arcos Radial ICP-OES. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001