Learn More
Glass Expansion Injector Adaptor for D-Torch Spectro EOP
SDP
Catalog No. 502703134
Encompass_Preferred
Injector Adaptor for D-Torch. Compatible with Glass Expansion’s D-Torch (501907029, 30-808-3371) for Spectro (Ametek) EOP Modula, Ciros, Genesis, and Arcos Axial ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3134 Supplier Glass Expansion Supplier No. 318083374
Injector Adaptor for D-Torch. Compatible with Glass Expansion’s D-Torch (501907029, 30-808-3371) for Spectro (Ametek) EOP Modula, Ciros, Genesis, and Arcos Axial ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001