Learn More
Glass Expansion Injector Adapter for D-Torch
SDP
Catalog No. NC2328923
Encompass_Preferred
Injector Adaptor for D-Torch. Compatible with Glass Expansion’s D-Torch Torch for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2328923 Supplier Glass Expansion Supplier No. 318082895
Injector Adaptor for D-Torch. Compatible with Glass Expansion’s D-Torch Torch for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001