Learn More
Perkin Elmer US LLC Injector Adapter Kit for NexION 300/350
Catalog No. NC1777947
Encompass_Preferred
This kit includes: O-Ring 6.07 mm I.D., Qty. 1 (09210011) O-Ring 14.0 mm I.D., Qty. 1 (N8122010) O-Ring 9.53 mm I.D., Qty. 1 (W1008572) O-Ring 15.9 mm I.D., Qty. 1 (W1013545) Polycarbonate Torch Adapter, Qty. 1 (W1012406) Injector Nut, Qty. 1 (W1012408) Injector Spacer, Qty. 1 (W1012409)
Catalog No. NC1777947 Supplier Perkin Elmer US LLC Supplier No. N8140527
This kit includes: O-Ring 6.07 mm I.D., Qty. 1 (09210011) O-Ring 14.0 mm I.D., Qty. 1 (N8122010) O-Ring 9.53 mm I.D., Qty. 1 (W1008572) O-Ring 15.9 mm I.D., Qty. 1 (W1013545) Polycarbonate Torch Adapter, Qty. 1 (W1012406) Injector Nut, Qty. 1 (W1012408) Injector Spacer, Qty. 1 (W1012409)