Learn More
Perkin Elmer US LLC Alumina Injector, 0.8 mm I.D.
Catalog No. 509043887
Encompass_Preferred
This Alumina Injector is for use with Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.
Catalog No. 50-904-3887 Supplier Perkin Elmer US LLC Supplier No. N0775227
This Alumina Injector is for use with Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.