Learn More
Ted Pella Inc Injector Blades, 20 blades in dispenser
SDP
Supplier:  Ted Pella Inc 1214
Encompass
Acceptable for use with Microslicer™, Vibratome and other vibratory microtomes.Single edge, 20 per dispenser.
Catalog No. NC1541673