Learn More
Waters Corp Injector Cartridge, FTN, 18 kpsi, Assembly
Catalog No. 502390853 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This injector cartridge is designed for high-pressure liquid chromatography applications, capable of operating up to 18,000 psi. It serves as a critical component in the fluidic pathway of compatible systems, ensuring precise and repeatable sample introduction.
  • Designed for high-pressure liquid chromatography applications.
  • Operates at pressures up to 18,000 psi.
  • Functions as a critical component in the fluidic pathway.
  • Ensures precise and repeatable sample delivery.
  • Helps maintain system integrity in demanding conditions.
  • Replaced during routine maintenance.
  • Useful for addressing flow path blockages or pressure issues.
  • Compatible with specific UPLC systems.
Catalog No. 50-239-0853 Supplier Waters Corp Supplier No. 700006057
This injector cartridge is designed for high-pressure liquid chromatography applications, capable of operating up to 18,000 psi. It serves as a critical component in the fluidic pathway of compatible systems, ensuring precise and repeatable sample introduction.
  • Designed for high-pressure liquid chromatography applications.
  • Operates at pressures up to 18,000 psi.
  • Functions as a critical component in the fluidic pathway.
  • Ensures precise and repeatable sample delivery.
  • Helps maintain system integrity in demanding conditions.
  • Replaced during routine maintenance.
  • Useful for addressing flow path blockages or pressure issues.
  • Compatible with specific UPLC systems.