Learn More
Glass Expansion Injector Ferrule Tool - 6.5 mm
SDP
Catalog No. 502703078
Encompass_Preferred
Injector Ferrule Tool - 6.5 mm. Provided with Glass Expansion D-Torches to facilitate the removal and installation of the injector ferrule. ONLY compatible with Injector Ferrule P/N 31-808-2897 for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES D-Torches. NOT compatible with Injector Ferrule (31-808-2962) for all other D-Torches. The injector ferrule is a consumable part that needs to be replaced on average about every 6 months. This tool should be used to remove and install the Injector Ferrule. Replacing the injector ferrule on a routine basis ensures a proper injector seal for optimum analytical performance.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3078 Supplier Glass Expansion Supplier No. 708030915
Injector Ferrule Tool - 6.5 mm. Provided with Glass Expansion D-Torches to facilitate the removal and installation of the injector ferrule. ONLY compatible with Injector Ferrule P/N 31-808-2897 for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES D-Torches. NOT compatible with Injector Ferrule (31-808-2962) for all other D-Torches. The injector ferrule is a consumable part that needs to be replaced on average about every 6 months. This tool should be used to remove and install the Injector Ferrule. Replacing the injector ferrule on a routine basis ensures a proper injector seal for optimum analytical performance.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001