Learn More
Perkin Elmer US LLC Sapphire Injector for Demountable ZipTorch, 1.8 mm I.D. Optima 4/5/7x00
Catalog No. 509047634
Encompass_Preferred
This Demountable Injector is for use with Optima 4x00/5x00/7x00 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.
Catalog No. 50-904-7634 Supplier Perkin Elmer US LLC Supplier No. N0777044
This Demountable Injector is for use with Optima 4x00/5x00/7x00 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.