Learn More
Perkin Elmer US LLC Alumina Injector, 1.2 mm I.D. - Entire Length of Tube for Optima 2/4/5/7x00
Catalog No. 509043920
Encompass_Preferred
This Alumina Injector is for use with Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.
Catalog No. 50-904-3920 Supplier Perkin Elmer US LLC Supplier No. N0776093
This Alumina Injector is for use with Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.