Learn More
Trajan Scientific INJECTOR SAMPLE 10F-GT
SDP
Catalog No. 502530402
Encompass_Preferred
SGE Gastight Syringes: Fixed Needle Models, Volume: 10 uL, Needle Gauge: 26 G, Model: 10F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm, For Use With: Suitable for gases or liquids for maximum inertness
Catalog No. 50-253-0402 Supplier Trajan Scientific Supplier No. 002200
SGE Gastight Syringes: Fixed Needle Models, Volume: 10 uL, Needle Gauge: 26 G, Model: 10F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm, For Use With: Suitable for gases or liquids for maximum inertness