Learn More
Trajan Scientific INJECTOR SAMPLE 1BR-5
SDP
Catalog No. 502530428
Encompass_Preferred
SGE NanoVolume Syringes, Volume: 1 uL, Needle Gauge: 23 G, Model: 1BR-5, Needle Point Style: Removable Cone, Length Needle: 50 mm, 1.96 in., Accuracy: +/-2%, Connection Type: Removable, Diameter Outer: 0.315 in., 8 mm, Diameter Outer Needle: 0.0248 in., 0.63 mm, Length: 2 in., 50 mm
Catalog No. 50-253-0428 Supplier Trajan Scientific Supplier No. 000500
SGE NanoVolume Syringes, Volume: 1 uL, Needle Gauge: 23 G, Model: 1BR-5, Needle Point Style: Removable Cone, Length Needle: 50 mm, 1.96 in., Accuracy: +/-2%, Connection Type: Removable, Diameter Outer: 0.315 in., 8 mm, Diameter Outer Needle: 0.0248 in., 0.63 mm, Length: 2 in., 50 mm