Learn More
Perkin Elmer US LLC Injector Septum Cap Assembly
Catalog No. 502117316
Encompass_Preferred
The injector septum assembly is compatible with the Capillary Split/Splitless (Cap), Packed, Temperature Programmable Spilt/Splitless (PSS), Temperature Programmable On-Column (POC) injectors for the Clarus 500/580 and Clarus 600/680 Gas Chromatographs. Note that this cap is septum cap is not compatible with the Clarus 590/690 capillary injector.
Catalog No. 50-211-7316 Supplier Perkin Elmer US LLC Supplier No. N6100153
The injector septum assembly is compatible with the Capillary Split/Splitless (Cap), Packed, Temperature Programmable Spilt/Splitless (PSS), Temperature Programmable On-Column (POC) injectors for the Clarus 500/580 and Clarus 600/680 Gas Chromatographs. Note that this cap is septum cap is not compatible with the Clarus 590/690 capillary injector.