Learn More
Molecular Devices Molecular Devices Microplate Reader, Multi-Mode, Tunable Monochromator, 6-384 Well
Catalog No. 5000014373
Encompass_Preferred
The SpectraMax iD3 multi-mode microplate reader from Molecular Devices provides absorbance, fluorescence (top and bottom), and luminescence detection for 6-384 well microplates. It features a tunable monochromator with 1-nm increments, intuitive touchscreen control, and software tools for spectral scanning and assay optimization, enabling rapid assay development and reliable quantitative measurements.
  • Multi-mode detection for absorbance, fluorescence, and luminescence assays.
  • Tunable monochromator with 1-nm wavelength increments for spectral flexibility.
  • Broad wavelength coverage for diverse assay types, including UV and visible ranges.
  • Supports 6 to 384 well plate formats for assay throughput flexibility.
  • Intuitive touchscreen and software tools for quick setup and assay optimization.
  • High photometric precision and repeatability for reliable quantitative results.
This item is not returnable. View return policy
Catalog No. 50-000-14373 Supplier Molecular Devices Supplier No. ID3STD
Add to Cart
Add to Cart
Request a Quote
The SpectraMax iD3 multi-mode microplate reader from Molecular Devices provides absorbance, fluorescence (top and bottom), and luminescence detection for 6-384 well microplates. It features a tunable monochromator with 1-nm increments, intuitive touchscreen control, and software tools for spectral scanning and assay optimization, enabling rapid assay development and reliable quantitative measurements.
  • Multi-mode detection for absorbance, fluorescence, and luminescence assays.
  • Tunable monochromator with 1-nm wavelength increments for spectral flexibility.
  • Broad wavelength coverage for diverse assay types, including UV and visible ranges.
  • Supports 6 to 384 well plate formats for assay throughput flexibility.
  • Intuitive touchscreen and software tools for quick setup and assay optimization.
  • High photometric precision and repeatability for reliable quantitative results.