Learn More
Waters Corp Waters Injector Valve Cartridge, SM-FTN, for ACQUITY Systems, 1 pk
Catalog No. 50793079 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement injector valve cartridge is designed for use on Flow Through Needle (FTN) Sample Managers, Patrol Sample Manager, and HDX Manager within various liquid chromatography systems.
  • Replacement injector valve cartridge.
  • For use on Flow Through Needle (FTN) Sample Managers.
  • Compatible with Patrol Sample Manager.
  • Compatible with HDX Manager.
  • Performance maintenance kit.
  • Includes 1 unit per package.
  • Designed for HPLC and UPLC system types.
  • Supports instruments such as iS HPLC System, Arc HPLC System, UPLC H-Class System, and UPLC M-Class System.
  • Component for Arc Sample Manager FTN-R.
  • Component for UPLC Process Sample Manager.
  • Replacement is recommended as required.
Catalog No. 50-793-079 Supplier Waters Corp Supplier No. 700005236
This replacement injector valve cartridge is designed for use on Flow Through Needle (FTN) Sample Managers, Patrol Sample Manager, and HDX Manager within various liquid chromatography systems.
  • Replacement injector valve cartridge.
  • For use on Flow Through Needle (FTN) Sample Managers.
  • Compatible with Patrol Sample Manager.
  • Compatible with HDX Manager.
  • Performance maintenance kit.
  • Includes 1 unit per package.
  • Designed for HPLC and UPLC system types.
  • Supports instruments such as iS HPLC System, Arc HPLC System, UPLC H-Class System, and UPLC M-Class System.
  • Component for Arc Sample Manager FTN-R.
  • Component for UPLC Process Sample Manager.
  • Replacement is recommended as required.