Learn More
Instech Injectors to manually access the ports in Instech VAH95AB Vascular Access Harnesses, VAB95BS buttons and SIP22/4 injection ports. Needlefree. 10 sterile pouches of 5. Replaces older SN22 needles.
SDP
Catalog No. NC0658135
Encompass
The Instech VAH6M-50 injector provides needle-free, protected access to rat Vascular Access Harness™ ports for flushing, dosing, or sampling. Its recessed tip interfaces safely with the VAH septum while preventing needle-stick risk. Compatible with rat VAH and VAB95 systems, it supports quick manual access during infusion or blood-collection workflows. Supplied as 50 sterile pouches of 5 for high-throughput studies.
Features
  • Injector for rat VAH™ and VAB95 systems
  • Recessed, needle-free protected tip
  • For manual flush, dose, or sampling
  • 50 sterile pouches of 5
  • Compatible with 3Fr rat catheter setups
  • EtO sterilized
Catalog No. NC0658135 Supplier Instech Supplier No. VAH6M50
The Instech VAH6M-50 injector provides needle-free, protected access to rat Vascular Access Harness™ ports for flushing, dosing, or sampling. Its recessed tip interfaces safely with the VAH septum while preventing needle-stick risk. Compatible with rat VAH and VAB95 systems, it supports quick manual access during infusion or blood-collection workflows. Supplied as 50 sterile pouches of 5 for high-throughput studies.
Features
  • Injector for rat VAH™ and VAB95 systems
  • Recessed, needle-free protected tip
  • For manual flush, dose, or sampling
  • 50 sterile pouches of 5
  • Compatible with 3Fr rat catheter setups
  • EtO sterilized