Learn More
Medchemexpress LLC HY-13328 100mg , INK-128 Sapanisertib;MLN0128 CAS:1224844-38-5 Purity:98%
SDP
Catalog No. 501492672
Encompass_Preferred
Medchemexpress, HY-13328 100mg INK-128 Sapanisertib;MLN0128 CAS:1224844-38-5 Formula:C15H15N7O IC50: 1 nM (mTOR), 219 nM (PI3Kα), 5293 nM (PI3Kβ), 230 nM (PI3Kδ), 221 nM (PI3Kγ) Ki: 1.4 nM (mTOR), 152 nM (PI3Kα), 4700 nM (PI3Kβ), 165 nM (PI3Kγ) Purity:98% INK-128 is a potent and selective  mTOR  inhibitor with  IC 50  of 1 nM, > 200-fold less potent to class I PI3K isoforms, superior in blocking mTORC1 / 2 and sensitive to pro-invasion genes. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2672 Supplier Medchemexpress LLC Supplier No. HY13328100MG
Medchemexpress, HY-13328 100mg INK-128 Sapanisertib;MLN0128 CAS:1224844-38-5 Formula:C15H15N7O IC50: 1 nM (mTOR), 219 nM (PI3Kα), 5293 nM (PI3Kβ), 230 nM (PI3Kδ), 221 nM (PI3Kγ) Ki: 1.4 nM (mTOR), 152 nM (PI3Kα), 4700 nM (PI3Kβ), 165 nM (PI3Kγ) Purity:98% INK-128 is a potent and selective  mTOR  inhibitor with  IC 50  of 1 nM, > 200-fold less potent to class I PI3K isoforms, superior in blocking mTORC1 / 2 and sensitive to pro-invasion genes. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.