Learn More
Apexbio Technology LLC INK 128 (MLN0128) 1224844-38-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A85515.1
Encompass_Preferred
INK 128 (MLN0128 CAS 1224844-38-5) is a selective inhibitor of the mammalian target of rapamycin (mTOR) a conserved serine/threonine kinase integral to the PI3K/AKT/mTOR signaling pathway This compound inhibits both mTOR complexes (mTORC1 and mTORC2) demonstrating an IC50 of approximately 1 nM In preclinical studies utilizing human pancreatic and HER2-positive breast cancer cell lines INK 128 significantly reduced cellular proliferation and viability via concentration- and time-dependent suppression of mTOR activity Moreover it exhibited notable tumor suppression in breast cancer xenograft models enhancing efficacy when combined with agents such as sorafenib sunitinib paclitaxel or lapatinib highlighting its potential value for oncology research and therapeutic development
Catalog No. 50-000-61697