Learn More
Selleck Chemical LLC Sapanisertib (MLN0128) 5mg 1224844-38-5 INK 128,TAK-228
SDP
Supplier:  Selleck Chemical LLC S28115MG
Encompass_Preferred
Sapanisertib (MLN0128, INK 128, TAK-228) is a potent and selective mTOR inhibitor with IC50 of 1 nM in cell-free assays; >200-fold less potent to class I PI3K isoforms, superior in blocking mTORC1/2 and sensitive to pro-invasion genes (vs Rapamycin). Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0943011