Learn More
GA International Ink Ribbon Takeup Core for 4″ – Wide Class Thermal-Transfer Printers (Carton), (Pack of 3) 3 cores/pack
Catalog No. NC3361139
Encompass_Preferred
Width: 4.33″ (110 mm), Inner Diameter: 1.0″ (25.4 mm), Material: Carton. Pack of three carton ink ribbon takeup cores for 4″ wide-class of desktop and industrial thermal-transfer printers such as Zebra, cab, Sato, Toshiba, and GA International. Accepts up to 4.33″ wide ink ribbons and fits most standard 1″ diameter spindles or core holders.
Catalog No. NC3361139 Supplier GA International Supplier No. C1C4.333
Width: 4.33″ (110 mm), Inner Diameter: 1.0″ (25.4 mm), Material: Carton. Pack of three carton ink ribbon takeup cores for 4″ wide-class of desktop and industrial thermal-transfer printers such as Zebra, cab, Sato, Toshiba, and GA International. Accepts up to 4.33″ wide ink ribbons and fits most standard 1″ diameter spindles or core holders.