Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TerraCycle™ Ink and Toner Cartridges Zero Waste Box™
SDPSCI_ED

Catalog No. S000934
Change view
Click to view available options
Capacity (Metric):
39.66 L
136.42 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000934 39.66 L
S000925 136.42 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options

These boxes are not suitable for medical waste. This includes any ‘red bag waste’ (i.e. material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians’ offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.) The zero waste box system does not accept household items that may be considered hazardous waste. Do not ship dripping packages. Not available in Hawaii, Alaska, and Puerto Rico.

Catalog No. S000934 Supplier TerraCycle™ Supplier No. CS80S
Add to Cart
Add to Cart

These boxes are not suitable for medical waste. This includes any ‘red bag waste’ (i.e. material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians’ offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.) The zero waste box system does not accept household items that may be considered hazardous waste. Do not ship dripping packages. Not available in Hawaii, Alaska, and Puerto Rico.

Recycle ink and toner cartridges. Each box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

  • What can you recycle?
    • Use this Zero Waste Box to recycle any brand and size of used ink and toner cartridge from personal or commercial printers. Do not send in printers, printer parts, or printer accessories. Do not send hazardous materials.
    • Do not send in medical waste. This includes material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians’ offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • How It Works
    • Order: Select a Zero Waste Box and place your order.
    • Collect: Collect the accepted items to fill the box.
    • Ship: Send it back to TerraCycle with the prepaid return label on your box.
    • Reorder: Order your next Zero Waste Box to continue recycling everything.
  • What’s Included?
    • One Zero Waste Box
    • One prepaid return label, affixed to your box
    • Processing and recycling of the materials
  • Important Shipping Restrictions
    • Only send items accepted by this box. Sending items outside the selected category causes contamination and added separation costs.
    • Do not send household items considered hazardous, including lithium-ion batteries, pressurized canisters, pesticides, oil-based paint, medical sharps, organics, broken glass, biomedical waste, soiled diapers, or expired medication. Avoid anything sharp, flammable, reactive, corrosive, ignitable, radioactive, toxic, infectious, or pathogenic.
    • Do not ship dripping packages. Remove excess product and ensure the box is completely dry before shipping.

Specifications

Capacity (Metric) 39.66 L
Capacity (English) 10.47 gal.
Color Brown
Dimensions (L x W x H) 11 x 11 x 20 in.
Autoclavable Not Autoclavable
Material Cardboard
Product Type Recycling Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.