Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Inkjet Printable Repositionable Sign Sheets

Catalog No. 19129948 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-948 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-948 Supplier Brady™ Supplier No. 151578
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Repositionable inkjet sign sheets create durable signage for areas where identification changes frequently

  • Inkjet-printable vinyl sheets support custom sign creation.
  • Repositionable adhesive allows signs to be moved without leaving residue.
  • Flexible durable material supports frequently changed identification.

Specifications

Adhesive Type Repositionable
Coating None
Color White
Finish Matte
Format Sheet
Material Vinyl
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) COVID-19, Facility Identification, Hazard Communication Identification
For Use With (Equipment) Ink Jet Printers
Labels per Sheet 1
No. per Pack 50/Pk.
Temperature Range (English) -40° to +60°C
Thickness (English) 0.0591 in.
Length (English) 11 in.
Width (English) 8.5 in.
Height (English) 11 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.