Learn More
Fisherbrand™ Inkless Chart Recorders for Isotemp Ultra-Low Temperature Chest Freezers
Catalog No. p-7227922
Change view
Click to view available options
For Use With (Equipment):
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
332166G11 Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
332791G11 Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 332166G11 Supplier Fisherbrand™ Supplier No. 332166G11
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Record seven days of temperature performance at temperatures to -150°C using Fisherbrand™ Isotemp Chart Recorders for Ultra-Low Temperature Freezers.
  • Seven-day recorder
  • Functions to -150°C
  • Factory installed
TRUSTED_SUSTAINABILITY
Specifications
Description Factory Installed
Type Chart Recorder
Temperature Range -150°C
For Use With (Equipment) Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Static Holding Time 7 Days