Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Cold samples: can measure down to -30°C
- TRIS compatible
Accessories & Care
- Immovable glass sleeve junction, refillable with FRISCOLYT-B™ (51343185)
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Recommended Storage
InLab storage solution
Specifications
Specifications
| Connector Types | Multi-Pin |
| Junction | Immovable Glass Sleeve |
| Material | Glass |
| pH Range | 1 to 11 |
| Type | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology |
| For Use With (Application) | Measuring samples at low temperatures or with low ion concentrations |
| For Use With (Equipment) | Multi-Pin to BNC/RCA (30281896) |
| Internal Reference | ARGENTHAL™ with Ag+-trap |
| Membrane | LoT Glass |
| Model | InLab™ Cool Pro-ISM |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?