Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Cool pH Electrodes

Catalog No. p-9390001
Click to view available options
Connector Types:
Multi-Pin
S7

A glass pH electrode with an integrated temperature sensor rated to -30°C, making it ideal for measuring cold samples.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Cold samples: can measure down to -30°C
  • TRIS compatible

Accessories & Care

  • Immovable glass sleeve junction, refillable with FRISCOLYT-B™ (51343185)
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH Electrode InLab Cool
Connector Types S7
Junction Immovable Glass Sleeve
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating -30°C to +80°C
Type Combination Electrode
Filling Solution Type FRISCOLYT-B
For Use With (Application) Samples down to a temperature of minus thirty degrees centigrade
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL with Ag+ trap
Membrane LoT Glass
Model InLab™ Cool
Response Time <20 sec.
Storage Requirements FRISCOLYT-B
Temperature (English) Operating -22°F to +176°F
Temperature (Metric) Range -30°C to +80°C
Length (English) 4.72 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.