Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Easy pH Electrodes

Catalog No. 01911486 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
BNC
S7
Model:
InLab™ Easy
InLab™ Easy BNC
For Use With (Equipment):
Fixed Cable to sensor
S7 to BNC (30281915)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Model For Use With (Equipment)
01-911-486 BNC InLab™ Easy BNC Fixed Cable to sensor
01-911-485 S7 InLab™ Easy S7 to BNC (30281915)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-911-486 Supplier METTLER TOLEDO™ Supplier No. 51343011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A durable, polysulfone pH electrode with an integral bulb protector and gel electrolyte to avoid the inconvenience of refilling in field. Temperature sensors sold separately.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Aquarium water
  • Soft drinks
  • Beer
  • General lab

Accessories & Care

  • Ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
  • Temperature sensors sold separately (NTC 30 kΩ: 51300164, PT1000: 51300165)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC
Junction Ceramic
Material Polysulfone
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode
Filling Solution Type Gel
For Use With (Application) Low maintenance, field measurement and educational use
For Use With (Equipment) Fixed Cable to sensor
Internal Reference Ag/AgCl
Membrane U Glass
Model InLab™ Easy BNC
Probe NTC 30 kilohm
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Range 0°C to 80°C
Length (English) 4.72 in.
Length (Metric) 120 mm
Length (Metric) Cable 1.2 m
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.