Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Expert pH Electrodes
GSA_VA

Catalog No. 01911461 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
BNC, RCA
BNC, RCA IP67
Multi-Pin
S7
Includes:
Electrode only
ISM Temperature Probe
Pt1000 Temperature Probe
Temperature Probe
Temperature Probe (NTC30)
Length (Metric) Cable:
1.2 m
1.8 m
10 m
2 m
No Cable
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Includes Length (Metric) Cable
01-911-461 BNC, RCA Temperature Probe 1.2 m
01-911-463 Multi-Pin Temperature Probe (NTC30) No Cable
01-911-460 S7 Electrode only No Cable
01-911-465 BNC, RCA ISM Temperature Probe 2 m
01-911-464 Multi-Pin Pt1000 Temperature Probe No Cable
01-910-552 BNC, RCA IP67 Temperature Probe 2 m
01-917-140 BNC, RCA IP67 ISM Temperature Probe 1.8 m
01-917-141 BNC, RCA IP67 ISM Temperature Probe 10 m
01-915-108 BNC, RCA ISM Temperature Probe 1.2 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 01-911-461 Supplier METTLER TOLEDO™ Supplier No. 51343101
Only null left
Add to Cart
Add to Cart

A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.

Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible

Accessories & Care

  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC, RCA
Junction Dual
Material PEEK
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode, Integrated Temperature Probe
Filling Solution Type Xerolyt Polymer
For Use With (Application) Emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides
For Use With (Equipment) Fixed Cable to sensor
Includes Temperature Probe
Internal Reference ARGENTHAL
Membrane U Glass
Model InLab Expert Pro
Probe NTC 30 kilohm ATC
Response Time <20 sec.
Storage Requirements FRISCOLYT-B
Temperature (English) Operating 0°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.7 in.
Length (English) Cable 47 in.
Length (Metric) 120 mm
Length (Metric) Cable 1.2 m
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.