Learn More
Mettler Toledo™ InLab™ OptiOx Pro-DES™ DO Calibration Tube
Catalog No. 01915860 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Calibration Sleeve
Catalog No. Type
01-915-860 Calibration Sleeve
1 options
Catalog No. 01-915-860 Supplier Mettler Toledo™ Supplier No. 30786582
Add to Cart
Add to Cart
Accessory for calibrating the dissolved oxygen sensor InLab OptiOx Pro-DES.
  • Accessory for calibrating the dissolved oxygen sensor InLab OptiOx Pro-DES.
  • For use with METTLER TOLEDO NineFocus and InLab OptiOx Pro-DES digital sensor
TRUSTED_SUSTAINABILITY
Specifications
Type Calibration Sleeve