Learn More
METTLER TOLEDO™ InLab™ pH Electrode Storage Solution
Catalog No. 01910276 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
01-910-276 250 mL
1 options
This item is not returnable. View return policy
Catalog No. 01-910-276 Supplier METTLER TOLEDO™ Supplier No. 30111142
Add to Cart
Add to Cart
This item is not returnable. View return policy
Recommended solution for storing InLab™ pH electrodes and replacing electrolyte in refillable pH electrodes
  • InLab™ pH Electrode Storage Solution provides optimal conditions to preserve pH probe sensors during the time between measurements and for short- or long-term storage
  • Probe electrolyte level should always be higher than the sample level
  • Routinely refilling or replacing the electrolyte solution is necessary to maintain good electrode performance
  • Using InLab™ pH electrodes along with InLab™ Storage Solution eliminates the risk of contaminating your samples with silver ions thanks to the ARGENTHAL™ reference system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) InLab™ pH Electrodes, Refillable pH Electrodes
Type Storage Solution
Quantity 250 mL