Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Power pH Electrodes
GSA_VA

Catalog No. 01917144 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
Multi-Pin
S7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-917-144 Multi-Pin
01-911-466 S7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-917-144 Supplier METTLER TOLEDO™ Supplier No. 51344211
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Corrosive acids and bases
  • Protein-rich samples
  • Dairy
  • High pH values
  • High temperature
  • Junction-clogging samples
  • Polymeric dispersions
  • TRIS compatible

Accessories & Care

  • Non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description pH Electrode InLab Power Pro-ISM
Connector Types Multi-Pin
Junction Ceramic
Material Glass
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 130°C
Type Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
Filling Solution Type DPA Gel
For Use With (Equipment) Multi-Pin to BNC/RCA (30281896)
Internal Reference SteadyForce
Membrane A41 Glass
Model InLab™ Power Pro-ISM
Probe NTC 30 kω
Storage Requirements 3 mol/L KCl
Temperature (English) Operating 32°F to 266°F
Temperature (Metric) Range 0°C to 130°C
Length (English) 6.7 in.
Length (Metric) 170 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.