Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Pure pH Electrodes

Catalog No. p-9433011 Shop All METTLER TOLEDO Products
Click to view available options
Connector Types:
Multi-Pin
S7

Catalog No. 01-917-146

Supplier: METTLER TOLEDO™ 51344172

Only null left
Add to Cart
Add to Cart

A glass pH electrode designed for ion-weak-samples such as ultrapure water and deionized water.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Pure water
  • Ultrapure water
  • Deionized water
  • Waste water
  • Highly saline solution, sea water
  • Low-ion strength

Accessories & Care

  • Immovable glass sleeve junction
  • LoT membrane glass
  • InLab Pure: refillable FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Pure Pro-ISM: refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH electrode InLab Pure Pro-ISM
Connector Types Multi-Pin
Junction Immovable Glass Sleeve
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode
Filling Solution Type 3 mol/L KCl Reference, 1mol/L KCl Bridge
For Use With (Application) Measuring samples at low temperatures or with low ion concentrations
For Use With (Equipment) Multi-Pin to BNC/RCA (30281896)
Includes ISM Temperature Probe
Internal Reference ARGENTHAL with Ag+ trap
Membrane LoT Glass
Model InLab™ Pure Pro-ISM
Probe NTC 30 kω
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Range 0°C to 80°C
Length (English) 6.7 in.
Length (Metric) 170 mm
Diameter (English) 0.47 in.
Diameter (Metric) Body 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.