Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Science pH Electrodes

Catalog No. 01911458 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
Multi-Pin
S7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-911-458 S7
01-917-145 Multi-Pin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-911-458 Supplier METTLER TOLEDO™ Supplier No. 51343070
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A pH electrode featuring a special glass body which is resistant to corrosive or organic samples, and is ideal for high temperatures and biological media. The high electrolyte outflow prevents contaminants from entering and clogging the junction, which would interfere with the reading.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Waste water
  • TRIS buffer
  • Micro-biological samples
  • Yeast fermentation solutions
  • Starch solutions
  • Organic solvents
  • General lab

Accessories & Care

  • Moveable sleeve junction
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types S7
Junction Moveable Glass Sleeve
Material Glass
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) Slurries and emulsions
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL with Ag+ trap
Membrane A41 Glass
Model InLab™ Science
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.7 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.