Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Science pH Electrodes

Catalog No. 01917145 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
Multi-Pin
S7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-917-145 Multi-Pin
01-911-458 S7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-917-145 Supplier METTLER TOLEDO™ Supplier No. 51344072
Add to Cart
Edge
Add to Cart

A pH electrode featuring a special glass body which is resistant to corrosive or organic samples, and is ideal for high temperatures and biological media. The high electrolyte outflow prevents contaminants from entering and clogging the junction, which would interfere with the reading.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Waste water
  • TRIS buffer
  • Micro-biological samples
  • Yeast fermentation solutions
  • Starch solutions
  • Organic solvents
  • General lab

Accessories & Care

  • Moveable sleeve junction
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types Multi-Pin
Junction Moveable Glass Sleeve
Material Glass
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode; Integrated Temperature Probe; Intelligent Sensor Management (ISM) Technology
Filling Solution Type 3mol/L KCl
For Use With (Equipment) Multi-Pin to BNC/RCA (30281896)
Includes ISM Temperature Probe
Internal Reference ARGENTHAL with Ag+ trap
Membrane A41 Glass
Model InLab™ Science Pro-ISM
Probe NTC 30 kilohm
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 6.7 in.
Length (Metric) 170 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.