Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Water Go pH Electrode

Catalog No. 01914310 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
BNC, RCA IP67
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-914-310 BNC, RCA IP67
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-914-310 Supplier METTLER TOLEDO™ Supplier No. 30253098
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A pH electrode that ensures your entire portable measuring system is able to withstand wet and demanding environments (IP67 for a water-proof measurement).

  • For use with METTLER TOLEDO Seven2Go and SevenGoDuo portable pH meters.
  • Capture electrode calibration and results with EasyDirect software.

Featured Applications

  • Low-ion strength (pure water)
  • Water
  • Field/Environmental sampling

Accessories & Care

  • Porous PTFE junction
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Recommended Storage

InLab storage solution

TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC, RCA IP67
Junction Porous PTFE
Material Polysulfone Shaft
pH Range 1 to 11
Type Combination Electrode; Integrated Temperature Probe (NTC 30 kΩ)
For Use With (Application) Measuring samples at low temperatures or with low ion concentrations
For Use With (Equipment) Fixed cable to sensor
Internal Reference ARGENTHAL™ with Ag+-trap
Membrane LoT Glass
Model InLab™ Water Go
Storage Requirements InLab storage solution
Temperature (Metric) Range 0°C to 80°C
Length (Metric) Stem 120 mm
Diameter (Metric) Body 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.