Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Solids pH Electrodes
GSA_VA

Catalog No. 01917143 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
BNC IP67
Multi-Pin
S7
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-917-143 Multi-Pin
01-911-468 S7
01-914-337 BNC IP67
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-917-143 Supplier METTLER TOLEDO™ Supplier No. 51344155
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A low maintenance glass pH electrode with an integrated temperature probe featuring a clog-free, open junction that eliminates the risk of protein fouling. Its spear shaped tip is designed for piercing solid and semi-solid food samples (IP67 rated).

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Meats
  • Cheese
  • Seafood
  • Dough
  • Milk and cream
  • Butter, yogurt, ice cream
  • Fruits and vegetables
  • Ready-made meals
  • Solid and semi-solid food samples

Accessories & Care

  • Open junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description pH Electrode InLab Solids Pro-ISM
Connector Types Multi-Pin
Junction Open
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
Filling Solution Type Xerolyt Extra Polymer
For Use With (Application) Measuring samples with a tough exterior, spear shaped tip has been designed for easy piercing of fruit, cheese, meats, etc.
For Use With (Equipment) Multi-Pin to BNC/RCA (30281896)
Internal Reference ARGENTHAL
Membrane LoT Glass
Model InLab™ Solids Pro-ISM
Probe NTC 30 kilohm
Response Time <20 sec.
Storage Requirements FRISCOLYT-B
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Range 0°C to 80°C
Tip Type Spear
Length (English) 0.98 in.
Length (Metric) 25 mm
Diameter (English) 0.23 in.
Diameter (Metric) 6 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.