Learn More
Waters Corp Oil, Inland 45, 1 Liter
Supplier:  Waters Corp 700004485
Encompass_Preferred
This Inland 45 oil is a Waters Quality part, ensuring compatibility and optimal performance in analytical instruments. It serves as pump oil, particularly suitable for systems employing rotary pumps, and is a key component in routine maintenance.
  • Designed for optimal performance in analytical instruments.
  • Functions as pump oil for systems using rotary pumps.
  • Utilized in maintenance procedures for instruments, including the Xevo G2 QTof/Tof.
  • Supports replacement of exhaust oil mist filter elements.
  • Facilitates oil changes in rotary pumps.
Catalog No. 50-793-010