Learn More
Chemglass Life Sciences Adapter, Inlet, 29/42 Inner Joint, 95mm Height
SDP
Catalog No. CG101202
Encompass_Preferred
Adapter, Inlet, 29/42 Inner Joint, 95mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG101202 Supplier Chemglass Life Sciences Supplier No. CG101202
Adapter, Inlet, 29/42 Inner Joint, 95mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.