Learn More
Chemglass Life Sciences Adapter, Inlet, Fritted Disc, 19/22 Inner Joint, 65mm Height
SDP
Supplier:  Chemglass Life Sciences CG101119
Encompass_Preferred
Adapter, Inlet, Fritted Disc, 19/22 Inner Joint, 65mm Height. Adapter has an extra coarse disc between the serrated hose connection and the inner joint. Fritted disc prevents solids from being introduced into the system. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG101119