Learn More
Chemglass Life Sciences Adapter, Inlet, Minum-Ware
SDP
Catalog No. MW3201
Encompass_Preferred
Adapter, Inlet, Minum-Ware. Used with thermometers, bleed tubes, etc. with outside diameters from 5-7mm. A vacuum tight seal is made using the supplied Viton O-ring and nylon compression cap while allowing the vertical adjustment of the thermometer or bleed tube. Supplied complete with compression cap (CG-352-01) and Viton O-ring (CG-305-107). Inner drip joint is a 14/10.
Catalog No. MW3201 Supplier Chemglass Life Sciences Supplier No. MW3201
Adapter, Inlet, Minum-Ware. Used with thermometers, bleed tubes, etc. with outside diameters from 5-7mm. A vacuum tight seal is made using the supplied Viton O-ring and nylon compression cap while allowing the vertical adjustment of the thermometer or bleed tube. Supplied complete with compression cap (CG-352-01) and Viton O-ring (CG-305-107). Inner drip joint is a 14/10.