Learn More
Chemglass Life Sciences Adapter, Inlet, Straight, 14/20 Joint Size, Hose Connection
SDP
Catalog No. CG106214
Encompass_Preferred
Adapter, Inlet, Straight, 14/20 Joint Size, Hose Connection. With serrated hose connection having an O.D. of 10mm at the largest serration sealed between the top outer and lower inner standard taper joints.
Catalog No. CG106214 Supplier Chemglass Life Sciences Supplier No. CG106214
Adapter, Inlet, Straight, 14/20 Joint Size, Hose Connection. With serrated hose connection having an O.D. of 10mm at the largest serration sealed between the top outer and lower inner standard taper joints.