Learn More
Grainger Fill-Rite Inlet Flange Kit, Cast Iron, 2 in, 1 ea
Catalog No. 502782324
Encompass_Preferred
The inlet flange kit is a essential replacement component designed to maintain the optimal performance of fuel transfer pumps. Constructed from durable cast iron, this kit provides a secure interface for pump installation and ensures reliable fluid handling within compatible systems.
  • Compatible with 1200, 2400, 4200, 4400, 600, SD1200, and SD600 series pumps
  • Durable cast iron material
  • Includes inlet flange, inlet screen, inlet gasket, and attaching bolts
  • Designed for 2 in. NPT male tank openings
  • Features 1 in. female suction port thread
Catalog No. 50-278-2324 Supplier Grainger Supplier No. 19NK05
The inlet flange kit is a essential replacement component designed to maintain the optimal performance of fuel transfer pumps. Constructed from durable cast iron, this kit provides a secure interface for pump installation and ensures reliable fluid handling within compatible systems.
  • Compatible with 1200, 2400, 4200, 4400, 600, SD1200, and SD600 series pumps
  • Durable cast iron material
  • Includes inlet flange, inlet screen, inlet gasket, and attaching bolts
  • Designed for 2 in. NPT male tank openings
  • Features 1 in. female suction port thread