Learn More
Radiometer America Radiometer ABL90 FLEX PLUS Inlet Connector Gasket
SDP
Catalog No. NC0114535
Encompass_Preferred
ABL90 FLEX PLUS Inlet Connector Gasket
Inlet Connector Gasket for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.
This item has a minimum qty of 2 per supplier requirements.
Catalog No. NC0114535 Supplier Radiometer America Supplier No. 834662
ABL90 FLEX PLUS Inlet Connector Gasket
Inlet Connector Gasket for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.
This item has a minimum qty of 2 per supplier requirements.