Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Inlet Feed Tubes
GREENER_CHOICESDP

Catalog No. 31500823 Shop All Kemtech Products
Change view
Click to view available options
For Use With:
C900200
C910200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
31-500-823 C900200
31-500-821 C910200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-823 Supplier Synthware™ Supplier No. C900001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with rotary evaporator condensers

Specifications

Autoclavable Autoclavable
Material Borosilicate Glass
For Use With C900200
For Use With (Application) Organic Synthesis
Color Clear
Product Type Inlet Feed Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.