Learn More
Waters Corp Waters Corporation Inlet Filter Assembly, 10-micron
Supplier:  Waters Corp 700000157
Encompass_Preferred
This inlet filter assembly is a replacement part designed for use on Reagent Managers. Located at the end of the solvent inlet tube, it provides 10-micron filtration to remove dirt and debris, significantly reducing solvent contamination and increasing the lifespan of the check valve.
  • Replacement 10-micron inlet filter assembly
  • Filters out dirt and debris
  • Significantly reduces solvent contamination
  • Increases the life of the check valve
  • Used on Reagent Managers
Catalog No. 50-792-318