Learn More
Agilent Technologies Agilent inlet liner, Ultra Inert, splitless, double taper. for GC
Catalog No. NC3453321
Encompass
Component Type- Inlet linerDeactivation- Ultra InertGlass Wool- NoneInjection Type- SplitlessInner Diameter (ID)- 4 mmLength- 78.5 mmLiner Geometry- Double taperOuter Diameter (OD)- 6.47 mmUNSPSC Code- 41115715Volume- 800 µL
Catalog No. NC3453321 Supplier Agilent Technologies Supplier No. 51903983
Component Type- Inlet linerDeactivation- Ultra InertGlass Wool- NoneInjection Type- SplitlessInner Diameter (ID)- 4 mmLength- 78.5 mmLiner Geometry- Double taperOuter Diameter (OD)- 6.47 mmUNSPSC Code- 41115715Volume- 800 µL