Learn More
Chemglass Life Sciences Adapter, Air Inlet/Outlet, M6
SDP
Catalog No. 501532112
Encompass
Adapter, Air Inlet/Outlet, 4mm Tubing to fit #16 Pump Tubing. Adapter can be used as a gas outlet pipe in lieu of a condenser in applications where the aeration rate or temperature is low or for short term batch applications. More often, this pipe is used to provide overlay aeration. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Thread size: M6. Hose Connection OD: 4mm. Fits Tubing Size 16. Replacement Silicone O-Ring: CLS-1705-010.
This item is not returnable. View return policy
Catalog No. 50-153-2112 Supplier Chemglass Life Sciences Supplier No. CLS138055
Adapter, Air Inlet/Outlet, 4mm Tubing to fit #16 Pump Tubing. Adapter can be used as a gas outlet pipe in lieu of a condenser in applications where the aeration rate or temperature is low or for short term batch applications. More often, this pipe is used to provide overlay aeration. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Thread size: M6. Hose Connection OD: 4mm. Fits Tubing Size 16. Replacement Silicone O-Ring: CLS-1705-010.