Learn More
Welch™ Inlet Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
Catalog No. 01184157
Change view
Click to view available options
For Use With:
Rotary Vane Pump
Catalog No. For Use With
01-184-157 Rotary Vane Pump
1 options
Catalog No. 01-184-157 Supplier Welch™ Supplier No. 320016
Add to Cart
Add to Cart
For use with CRVpro Direct Drive Rotary Vane Vacuum Pump
  • Intake separator separates liquid phase aerosols from gas stream, reducing chemical contaminants in pump oil
Specifications
Description Inlet Separator
For Use With (Application) Separates liquid phase aerosols from gas stream
For Use With Rotary Vane Pump